missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-CoV-2 Nucleocapsid Monoclonal Antibody (bcn14)

Human Monoclonal Antibody
Marca: Invitrogen™ MA535955
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Purity: Greater than 95% as determined by reducing SDS-PAGE.
SARS-CoV-2 Nucleocapsid Protein is associated with nucleic acid. It is the most abundant protein for coronavirus. Because of the strong immunogenicity of coronavirus Nucleocapsid, it is believed that SARS-CoV-2 Nucleocapsid Protein has potential value for the diagnosis of the virus. The SARS-CoV-2 N protein is an abundant RNA-binding protein critical for viral genome packaging, yet the molecular details that involve this process are poorly understood. N protein contains three dynamic disordered regions that house putative transiently-helical binding motifs. The two folded domains interact minimally such that full-length N protein is a flexible and multivalent RNA-binding protein. SARS-CoV-2 causes illnesses ranging from the common cold to severe diseases.
Especificaciones
| SARS-CoV-2 Nucleocapsid | |
| Monoclonal | |
| 0.42 mg/mL | |
| PBS with 0.02% sodium azide; pH 7.2 | |
| P0DTC9 | |
| N | |
| Fully humanized monoclonal antibody paired with human monoclonal antibody corresponding to SARS-CoV-2 Nucleocapsid. | |
| 100 μg | |
| Primary | |
| Virus | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| bcn14 | |
| Unconjugated | |
| N | |
| COVID19 Nucleocapsid; COVID-19 nucleocapsid; COVID19 Spike nucleocapsid; SARS-CoV-2 N | |
| Human | |
| Protein A | |
| RUO | |
| 43740575 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido