missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 B.1.351S Spike RBD Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB11024-100
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Spike RBD Monoclonal antibody specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for Immunocytochemistry
Especificaciones
| Spike RBD | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| Immunocytochemistry | |
| 1049541 | |
| Immunocytochemistry 8-25 ug/mL | |
| YP_009724390 | |
| Human embryonic kidney cell HEK293-derived SARS-CoV-2 B.1.351S Spike RBD, 16-1211, Accession # YP_009724390 | |
| 100 μg | |
| Primary | |
| SARS-CoV-2 | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido