missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS Coronavirus Spike Protein Polyclonal Antibody

Descripción
The immunogen is 69%(11/16) homologous to SARS-COV-2 (COVID-19) Spike protein. The cross reactivity is not empirically tested. Suggested positive control: transfected mouse melanoma cell lysate. Immunogen sequence: AVDCALDPLS ETKCTL.
It has recently been shown that SARS (severe acute respiratory syndrome) is caused by a human coronavirus. Human coronaviruses are the major cause of upper respiratory tract illness, such as the common cold, in humans. Coronaviruses are positive-stranded RNA viruses, featuring the largest viral RNA genomes known to date (27-31 kb). The first step in coronavirus infection is binding of the viral spike protein, a 139-kDa protein, to certain receptors on host cells. The spike protein is the main surface antigen of the coronavirus. The glycosylated spike protein (as well as the nucleocapsid protein) can be detected in infected cell culture supernatants with antisera from SARS patients.
Especificaciones
Especificaciones
| Antígeno | SARS Coronavirus Spike Protein |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1.0 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.2% gelatin and 0.05% sodium azide |
| Alias de gen | Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 |
| Especie del huésped | Rabbit |
| Inmunógeno | Synthetic peptide corresponding to amino acids 288-303 from the S (Spike glycoprotein) (Spike protein 1) for the Human SARS coronavirus. |
| Método de purificación | Protein G |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?