missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAR1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA568201
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Predicted to react with Mouse and Rat samples.
Involved in transport from the endoplasmic reticulum to the Golgi apparatus. Required to maintain SEC16A localization at discrete locations on the ER membrane perhaps by preventing its dissociation. SAR1A-GTP-dependent assembly of SEC16A on the ER membrane forms an organized scaffold defining endoplasmic reticulum exit sites (ERES). [UniProt].
Especificaciones
| SAR1A | |
| Polyclonal | |
| Unconjugated | |
| SAR1A | |
| 1600019H17Rik; COPII-associated small GTPase; GTP-binding protein SAR1a; masra2; SAR1; SAR1 gene homolog A; SAR1 gene homolog A (S. cerevisiae); SAR1 homolog A; SAR1A; SAR1a gene homolog 1; Sara; Sara1; secretion associated Ras related GTPase 1A; secretion associated, Ras related GTPase 1A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20224, 361842, 56681 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P36536, Q9NR31 | |
| SAR1A | |
| Synthesized peptide derived from human SAR1A. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido