Learn More
Skytec™ Sapphire Zinc Gloves
Descripción
- High Cut Resistant Uncoated Glove Made with Engineered HPPE
- Cut resistant: Level E / A9
- Engineered HPPE offers outstanding cut protection
- Glass and steel fiber free reduce risks of irritation
- Spandex in the liner allows the glove to fit the hand perfectly
- Designed for easy movement and continuous wear
- Breathable to reduce perspiration
- Seamless knit prevents irritation
- Elasticated wrist provides a secure fit
- Ambidextrous for easy and quick donning
Especificaciones
Especificaciones
| Certificaciones/conformidad (Europa) | CE II, UKCA, ANSI/ISEA 105-2016: Cut A5, EN 388:2016 + A1:2018 2X43E, EN 21420:2020 |
| Coating Material | Uncoated |
| Color | Gray |
| Cut Resistance Rating | Level E |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | HPPE |
| Tipo de producto | Cut-resistant Gloves |
| Tamaño | Pequeño |
| Norma EN388 | 2X43E |
| Mostrar más |