Learn More
Invitrogen™ SAP97 Recombinant Rabbit Monoclonal Antibody (24GB5305)
GREENER_CHOICE
Código de producto. 30275661
Change view
Click to view available options
Cantidad:
20 μL
100 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
30275661 100 μL 100 microlitros
30275541 20 μL 20 microlitros
2 options
Código de producto. 30275661
Proveedor: Invitrogen™ MA557017
Rabbit Recombinant Monoclonal Antibody
SAP97 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a multi-domain scaffolding protein that is required for normal development. This protein may have a role in septate junction formation, signal transduction, cell proliferation, synaptogenesis and lymphocyte activation. Several alternatively spliced transcript variants encoding different isoforms have been described for this gene, but the full-length nature of some of the variants is not known.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno SAP97
Aplicaciones Flow Cytometry, Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon 24GB5305
Concentración 0.62 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
génica DLG1
N.º de referencia del gen Q12959, Q62696, Q811D0
Alias de gen B130052P05Rik; discs large homolog 1; discs large homolog 1, scribble cell polarity complex component; discs large MAGUK scaffold protein 1; discs, large homolog 1; discs, large homolog 1 (Drosophila); discs, large homolog 1 (Drosophila) transcript variant 6-v1; discs, large homolog 1 (Drosophila) transcript variant 6-v2; disks large homolog 1; dJ1061C18.1.1; DKFZp761P0818; DKFZp781B0426; DLG1; Dlgh1; Drosophila discs-large tumor suppressor homologue (synapse associated protein); E-dlg/SAP97; Embryo-dlg/synapse-associated protein 97; hDlg; mKIAA4187; presynaptic protein SAP97; SAP97; SAP-97; synapse-associated protein 97
Símbolos de los genes DLG1
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from human SAP97 (aa 1-44).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 13383, 1739, 25252
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG