Descripción
Meet US and EU standards for use in pharmaceutical process applications. ASME-BPE, USP Class VI, and FDA compliant. Sterilize by autoclave, EtO, or gamma irradiation. Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application
Especificaciones
Especificaciones
| Certificaciones/conformidad | ADCF;FDA;Non-Phthalate;RoHS;USP |
| Temperatura mín. de funcionamiento (métrico) | 135°C |
| Puerto de salida | Sanitario |
| Método de esterilización | Autoclave, EtO, gamma irradiation |
| Incluye | 3/4 in Tamaño de la abrazadera sanitaria |