Learn More
Saint-Gobain Performance Plastics Sani-Tech™ STHT™-80 Tubing with 9.5 mm ID
Product Code. p-200132110
Change view
Click to view available options
Diámetro exterior (imperial):
0.500 in., 12,7 mm
0.563 in., 14,3 mm
0.625 in., 15,9 mm
Cantidad:
1 rollo
10 rollos
20 rollos
Unit Size:
150 metros
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diámetro exterior (imperial) Cantidad unitSize
30428833 0.563 in., 14,3 mm 20 rollos 150 metros
30428832 0.500 in., 12,7 mm 20 rollos 150 metros
30428826 0.625 in., 15,9 mm 20 rollos 150 metros
30428874 0.563 in., 14,3 mm 10 rollos 150 metros
30428807 0.500 in., 12,7 mm 1 rollo 150 metros
30428868 0.625 in., 15,9 mm 10 rollos 150 metros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30428833 Supplier Saint-Gobain Performance Plastics Supplier No. STHT803753
This item is not returnable. View return policy
Excels in high burst pressure and vacuum applications. With superior pressure resistance, it eliminates costly braid reinforcements. It reduces pulsation for accurate filling and dosing and its transparency ensures easy visual monitoring.
  • Sani-Tech™ STHT™-80 is a premium platinum-cured silicone tubing designed for high burst pressure and vacuum applications.
  • Offers superior pressure resistance, eliminating the need for costly braid reinforcements.
  • Ideal for fill finish operations, reducing pulsations downstream of pumping for precise filling and dosing accuracy.
  • Exhibits two to three times less pulsation compared to competing high-durometer tubings, helping prevent unintended dripping at the filling needle.
  • Achieves best-in-class performance with the highest burst pressure ratings among comparable products.
  • Maintains vacuum compatibility, making it suitable for a wide range of critical fluid transfer applications.
  • Offers high transparency for easy visual monitoring of fluid activity within the tubing, supporting applications that require visual cues and observation.
  • Supports consistent, accurate fluid transfer, enhancing the reliability of critical processes.
TRUSTED_SUSTAINABILITY
Specifications
Esterilizables en autoclave Autoclavable
Diámetro interior (imperial) 0.375 in.
Diámetro interior (métrico) 9.5 mm
Material Silicone
Diámetro exterior (imperial) 0.563 in., 14,3 mm
Longitud global (imperial) 25 ft.
Longitud global (métrico) 7.62 m
Color Translucent
Dureza 80 (Shore A)
Tipo de producto Pressure and Vacuum Tubing
Cantidad 20 rollos