missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: ALVRGTPVRG AITRGATVTR GVPPPPTVRG APAPRARTAG IQRIPLP Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a member of the FG-repeat containing nucleoporins that functions as a soluble cofactor in importin-alpha:beta-mediated nuclear protein import. Pseudogenes of this gene are found on chromosomes 5, 6, and 14. Two transcript variants encoding different isoforms have been found for this gene.
Especificaciones
Especificaciones
| Antígeno | SAM68 |
| Aplicaciones | ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | KHDRBS1 |
| N.º de referencia del gen | Q07666 |
| Alias de gen | FLJ34027; GAP-associated tyrosine phosphoprotein p62; GAP-associated tyrosine phosphoprotein p62 (Sam68); KH domain containing, RNA binding, signal transduction associated 1; KH domain-containing, RNA-binding, signal transduction-associated protein 1; KH RNA binding domain containing, signal transduction associated 1; KHDRBS1; khdrbs1 {ECO:0000312; p21 Ras GTPase-activating protein-associated p62; P62; p68; RGD:621459}; Sam68; src associated in mitosis, 68 kDa; src-associated in mitosis 68 kDa protein |
| Símbolos de los genes | KHDRBS1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
