Learn More
Macherey-Nagel™ SALTESMO Test Discs
Frequently used for semi-quantitative determination of salt (NaCl) in foodstuffs and in sea water. Macherey-Nagel™ SALTESMO Test Discs are impregnated papers that display a discolored zone proportional in size to the chloride concentration in the test solution.
Marca:  Macherey-Nagel™ 90608
Código de producto. 10340621
  • 97.70€ / paquete de 30
Solo quedan null
En stock
Descripción
All reagents are impregnated on the Saltesmo test disc paper, making the test very easy to use even for untrained persons. Simply pierce a hole into the disc using the needle supplied with the test and allow the disc to soak in the sample for 1-2 minutes. A discolored zone appears around the hole. The size of the colored zone is proportional to the chloride concentration in the solution.
  • Especially suitable for quick checks on halogen ion concentrations (NaCl, KCl, NaBr or KI)
  • Reacts with chloride, bromide and iodide, changing color from red-brown to yellow
  • Reference chart used to verify/measure concentration
Especificaciones
Especificaciones
Halide ions (NaCl, KCl, NaBr, KI)
Semi-quantitative Test Paper
0 - 0.25 - 0.5 - 1 - 2 - 3 - 4 - 5 g/L
30
0-5g/L
SALTESMO Test Discs
Red-brown to Yellow
For chloride, bromid or iodide determination (e.g. salt concentration in food or seawater)
Puncture needle, Reference scale, Box
30 Pack
1 to 2 min.