Learn More
Macherey-Nagel™ SALTESMO Test Discs
Frequently used for semi-quantitative determination of salt (NaCl) in foodstuffs and in sea water. Macherey-Nagel™ SALTESMO Test Discs are impregnated papers that display a discolored zone proportional in size to the chloride concentration in the test solution.
Marca: Macherey-Nagel™ 90608
Descripción
All reagents are impregnated on the Saltesmo test disc paper, making the test very easy to use even for untrained persons. Simply pierce a hole into the disc using the needle supplied with the test and allow the disc to soak in the sample for 1-2 minutes. A discolored zone appears around the hole. The size of the colored zone is proportional to the chloride concentration in the solution.
- Especially suitable for quick checks on halogen ion concentrations (NaCl, KCl, NaBr or KI)
- Reacts with chloride, bromide and iodide, changing color from red-brown to yellow
- Reference chart used to verify/measure concentration
Especificaciones
| Halide ions (NaCl, KCl, NaBr, KI) | |
| Semi-quantitative Test Paper | |
| 0 - 0.25 - 0.5 - 1 - 2 - 3 - 4 - 5 g/L | |
| 30 | |
| 0-5g/L | |
| SALTESMO Test Discs |
| Red-brown to Yellow | |
| For chloride, bromid or iodide determination (e.g. salt concentration in food or seawater) | |
| Puncture needle, Reference scale, Box | |
| 30 Pack | |
| 1 to 2 min. |