missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Salmonella typhimurium LPS Monoclonal Antibody (1E6)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA183451
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Mouse anti Salmonella typhimurium LPS antibody, clone 1E6 reacts with an unidentified LPS determinant of Salmonella typhimurium.
The genus Salmonella is a member of the family Enterobacteriaceae. The genus is composed of Gram negative bacilli that are facultative and flagellated (motile). Salmonellae possess 3 major surface antigens; the H or flagellar antigen (phase 1 and 2), the O or somatic antigen (part of the LPS moiety) and the Vi or capsular antigen (referred to as K in other Enterobacteriaceae). Salmonellae also possess the LPS endotoxin characteristic of Gram negative bacteria. This LPS is composed of an O polysaccharide (O antigen) an R core and the endotoxic inner Lipid A. Endotoxins evoke fever and can activate complement, kinin and clotting factors.
Especificaciones
| Salmonella typhimurium LPS | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with <0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
| ELISA, Immunocytochemistry | |
| 1E6 | |
| Unconjugated | |
| S. typhimurium; Salmonella LPS; Salmonellae | |
| Purified LPS from S. typhimurium. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido