Descripción
HexArmor™ brand products stand for exceptional protection.
- The exclusive SuperFabric™ brand material offers pinprick protection and extremely high cut protection.
- For exceptional impact protection, the patented IR-X™ Impact Exoskeleton™ is an attractive solution.
Spécification
Spécification
| Estructura | Seamless Knit |
| Coating Material | Polyurethane |
| Color | Blue, Light Blue |
| Cut Resistance Rating | ANSI A6 |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | SuperFabric™ |
| Tipo de producto | Cut-resistant Gloves |
| Tamaño | 6 |
| Sexo | Unisex |
| Afficher plus |