Learn More
S1PR1/EDG1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Código de producto. 16876595 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16876595 20 μL 20 microlitros
16866595 150 μL 150 microlitros
2 options
Código de producto. 16876595
Marca: Proteintech 551331AP20UL
Rabbit Polyclonal Antibody
The antibody is specific to S1PR1.
S1P1 is a G-protein coupled receptor for the bioactive lysosphingolipid sphingosine 1-phosphate (S1P) that is coupled to the G(i) subclass of heteromeric G proteins. S1P1 signaling leads to the activation of RAC1, SRC, PTK2/FAK1 and MAP kinases. It plays an important role in cell migration via its role in the reorganization of the actin cytoskeleton and formation of lamellipodia in response to stimuli that increase the activity of the sphingosine kinase SPHK1. S1P1 is required for normal chemotaxis toward sphingosine 1-phosphate, normal embryonic heart development, and normal cardiac morphogenesis.
Especificaciones
Antígeno S1PR1/EDG1
Aplicaciones Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.77 mg/mL
Conjugado Unconjugated
Formulación 0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3
génica S1PR1
N.º de referencia del gen O08530, P21453
Alias de gen CD363, CHEDG1, D1S3362, ECGF1, EDG 1, EDG1, S1P receptor 1, S1P receptor Edg 1, S1P1, S1PR1
Símbolos de los genes S1PR1
Especie del huésped Rabbit
Inmunógeno Peptide
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 13609, 1901
Especies diana Mouse, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG