Learn More
S100A8 Mouse anti-Human, Clone: CF-145, Abnova™
Mouse monoclonal antibody raised against native human S100A8.
Marca: Abnova MAB12905.50ug
Descripción
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and as a cytokine. Altered expression of this protein is associated with the disease cystic fibrosis. [provided by RefSeq]
Especificaciones
| S100A8 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2.5 ug/mL) Western Blot The optimal working dilution should be determined by the end user. | |
| 60B8AG/CAGA/CFAG/CGLA/CP-10/L1Ag/MA387/MIF/MRP8/NIF/P8 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 6279 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| CF-145 | |
| Mouse monoclonal antibody raised against native human S100A8. | |
| In PBS (0.02% sodium azide). | |
| S100A8 | |
| Native purified human granulocyte antigen. | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.