Learn More
Invitrogen™ RYK Monoclonal Antibody (F35 P7 D7 F5)
GREENER_CHOICE
Código de producto. 11554393
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
11554393 100 μg 100 microgramos
1 options
Código de producto. 11554393
Marca: Invitrogen™ MA1615
Mouse Monoclonal Antibody
This antibody detects RYK in human, mouse, and rat samples. MA1-615 has been successfully used in Western blot, procedures. By Western blot this antibody detects the RYK receptor-like tyrosine kinase-1 in cells. The MA1-615 immunogen corresponds to the ovalbumin-conjugated synthetic peptide C (462) L T E F H A A L G A Y V(475).
RYK is an atypical member of the family of growth factor receptor protein tyrosine kinases, differing from other members at a number of conserved residues in the activation and nucleotide binding domains. This gene product belongs to a subfamily whose members do not appear to be regulated by phosphorylation in the activation segment. It has been suggested that mediation of biological activity by recruitment of a signaling-competent auxiliary protein may occur through an as yet uncharacterized mechanism. A nine nucleotide insertion in some transcripts results in the SLG variant. It is not established whether this is a product of alternative splicing or a second gene, since evidence for a second gene or pseudogene on chromosome 17 exists.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno RYK
Aplicaciones Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon F35 P7 D7 F5
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1mg/mL BSA and 0.05% sodium azide
génica RYK
N.º de referencia del gen P34925, Q01887
Alias de gen AW536699; D3S3195; ERK3; ERK-3; growth factor receptor; hydroxyaryl-protein kinase; JTK5; JTK5A; JTK5A protein tyrosine kinase; kinase VIK; Met-related kinase; Mrk; NYK-R; receptor-like tyrosine kinase; RYK; RYK1; tyrosine-protein kinase RYK; Vik
Símbolos de los genes RYK
Especie del huésped Mouse
Inmunógeno Synthetic peptide corresponding to residues C(462) L T E F H A A L G A Y V(475) of RYK, conjugated to ovalbumin.
Método de purificación Protein G
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 140585, 20187, 6259
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1