missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUNX1 (middle) Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25315-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
RUNX1 belongs to the mammalian RUNX family of DNA binding proteins that regulate the expression of genes involved in cellular differentiation and cell cycle progression. RUNX1 is essential for hematopoietic stem cell generation in the vascular regions of the aorta, vitelline and umbilical arteries, yolk sac and placenta. RUNX1 is expressed in all tissues except brain and heart, with highest levels in thymus, bone marrow and peripheral blood.Especificaciones
| RUNX1 (middle) | |
| Polyclonal | |
| Unconjugated | |
| RUNX1 | |
| AML1, AML1 EVI 1, AMLCR1, CBF alpha 2, CBFA2, EVI 1, Oncogene AML 1, PEA2 alpha B, PEBP2 alpha B, PEBP2aB, RUNX1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12394, 861 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q01196, Q03347 | |
| RUNX1 | |
| RUNX1 (middle) Fusion Protein Ag17838 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido