missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
NogoA is a member of a family of integral membrane proteins termed reticulons that are thought to be involved in numerous disorders including neurodegenerative diseases. Reticulon proteins are known to regulate many cellular processes and interact with multiple proteins and receptors such as BACE. NogoA was initially identified as a myelin-associated neurite outgrowth inhibitor. It is highly expressed in oligodendrocytes in the white matter of the CNS, blocking its activity with antibodies or other factors results in improved axon regrowth and functional recovery in experimental CNS lesion models. NogoA has also been suggested to play a role in neurodegenerative diseases such as Amyotrophic lateral sclerosis, in which case NogoA is found at elevated levels in postmortem muscular samples, and multiple sclerosis (MS), in which case autoantibodies to NogoA have been found in serum and cerebrospinal fluid in MS patients.
Especificaciones
Especificaciones
| Antígeno | RTN4/NOGO |
| Aplicaciones | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.13 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | RTN4 |
| N.º de referencia del gen | Q99P72, Q9NQC3 |
| Alias de gen | ASY, Foocen, KIAA0886, Nbla00271, Nbla10545, Neurite outgrowth inhibitor, NI220/250, NOGO, NOGO A, Nogo B, Nogo C, Nogo protein, NOGOA, NOGOC, NSP, NSP CL, reticulon 4, Reticulon 5, RTN X, RTN4, RTN4 A, RTN4 B1, RTN4 B2, RTN4 C, SP1507 |
| Símbolos de los genes | RTN4 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?