missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RTN3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA553359
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: VPQVKQQTDK SSDCITKTTG LDMSEYNSEI PVVNLKTSTH QKTPVCSIDG STPITKSTGD WAEASLQQEN AITGKPVPDS LNSTKEFSIK GVQGNMQKQD DTLAELPGSP PEKCDSLGSG VATVKVVLPD DHLKDEMDW Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 48%.
RTN3 is a member of reticulons, which are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. Alternatively spliced transcript variants encoding different isoforms have been identified. Multiple promoters rather than alternative splicing of internal exons seem to be involved in this diversity.
Especificaciones
| RTN3 | |
| Polyclonal | |
| Unconjugated | |
| RTN3 | |
| ASY interacting protein; ASYIP; HAP; Homolog of ASY protein; neuroendocrine-specific protein-like 2; Neuroendocrine-specific protein-like II; nsp-c; NSPL2; NSPLII; Nsp-like II; NSP-like protein 2; NSP-like protein II; reticulon 3; reticulon-3; RTN3; RTN3-A1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10313 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95197 | |
| RTN3 | |
| Recombinant protein corresponding to Human RTN3. Recombinant protein control fragment (Product #RP-90845). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido