missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSV Monoclonal Antibody (2017/84)
Mouse Monoclonal Antibody
Marca: Invitrogen™ MA516467
Este artículo no se puede devolver.
Vea la política de devoluciones
Description
Mouse anti Respiratory Syncytial Virus antibody,clone 2017/84recognizes respiratory syncytial virus (RSV), a 100-350 nm enveloped virus belonging to the Paramyxoviridae, genus Pneumovirus, that causes minor respiratory infections in adults and bronchitis and bronchopneumonia in children.
Human respiratory syncytial virus (RSV) is a member of the Paramyxoviridae family, subfamily Pneumovirinae, that causes common respiratory tract infections such as measles and mumps. Its genome consists of single stranded, negative sense RNA that encodes three envelope glycoproteins, a small hydrophobic (SH) protein of unknown function, a major glycoprotein (G) known as the attachment protein, and a fusion (F) protein. Its name comes from the fact that F proteins on the surface of the virus cause the membranes on nearby cells to merge, forming syncytia. While RSV typically produces mild symptoms in adults, it is one of the leading causes of lower respiratory tract infections and hospital visits during infancy and childhood.
Spécification
| RSV | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 2017/84 | |
| Unconjugated | |
| human respiratory syncytial virus; RSV | |
| Native RSV. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu