missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS6KB1(phospho T421) & RPS6KB1 Protein Phosphorylation Antibody Pair, Abnova™
Antibody Pair
Marca: Abnova DP0215.1Set
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This protein phosphorylation antibody pair set comes with two antibodies, one against the RPS6KB1 protein, and the other against the specific T421 phosphorylated site of RPS6KB1 for use in in situ Proximity Ligation Assay.Especificaciones
| RPS6KB1 | |
| Unconjugated | |
| PS6K/S6K/S6K1/STK14A/p70(S6K)-alpha/p70-S6K/p70-alpha | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Phospho-RPS6KB1 T421 rabbit polyclonal antibody (20 ul) In PBS, 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. RPS6KB1 mouse monoclonal antibody (40 ug) In 1x PBS, pH 7.2 *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| In situ PLA | |
| ribosomal protein S6 kinase, 70kDa, polypeptide 1 | |
| RPS6KB1 | |
| RUO | |
| 6198 | |
| Human | |
| Ab Pair |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido