missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15198-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
WD repeat domain 5 (WDR5) is a member of the WD repeat protein family, which is involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. WDR5, also known as BIG-3, is expressed in the developing growth plate, accelerates chondrocyte and osteoblast differentiation in vitro, and regulates osteoblast differentiation during embryonic bone development. WDR5 interacts with the pluripotency factor Oct4/POU5F1 and is required for the efficient formation of induced pluripotent stem (iPS) cells.Especificaciones
| RPS3 | |
| Polyclonal | |
| Unconjugated | |
| RPS3 | |
| 40S ribosomal protein S3, ribosomal protein S3, RPS3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140654, 27050, 6188 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P23396, P62908, P62909 | |
| RPS3 | |
| RPS3 Fusion Protein Ag7346 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido