missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPN1 Monoclonal Antibody (OTI5A1), TrueMAB™, OriGene
Descripción
Ribophorins 1 and 2 are abundant and highly conserved glycoproteins residing in the endoplasic reticulum, that participate in ribosome binding. Mammalian oligosaccharyltransferase activity is associated with a protein complex composed of RPN1, RPN2, and an oligosaccharyltransferase protein. RPN1 is a component of the proteasome base. The ubiquitin-like (UBL) domain of recombit Rad23 interacts with proteasomes through the leucine-rich repeat domain of RPN1. The RPN1 gene maps to chromosome 3 in somatic cell hybrids, and the RPN2 gene maps to chromosome 20 by in situ hybridization.
Especificaciones
Especificaciones
| Antígeno | RPN1 |
| Aplicaciones | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI5A1 |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | RPN1 |
| N.º de referencia del gen | P04843 |
| Alias de gen | OST1, RBPH1 |
| Símbolos de los genes | RPN1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?