missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPLP0 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11290-2-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The protein encoded by this gene contains RNA recognition motif found in a variety of RNA binding proteins, including various hnRNP proteins, proteins implicated in regulation of alternative splicing, and protein components of snRNPs. In vitro studies showed that the rat homolog bound to RNA homopolymers, with a preference for G and U polyribonucleotides. This gene is part of a gene cluster on chromosome Xp11.23, and its 3′ end lies within 20 kb upstream of UBE1. Two transcript variants encoding different isoforms have been identified for this gene.Especificaciones
| RPLP0 | |
| Polyclonal | |
| Unconjugated | |
| RPLP0 | |
| 60S ribosomal protein L10E, L10E, P0, PRLP0, ribosomal protein, large, P0, RPLP0, RPP0 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11837, 6175, 64205 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P05388, P14869, P19945 | |
| RPLP0 | |
| RPLP0 Fusion Protein Ag1829 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido