missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: SVYAHFPINV VIQENGSLVE IRNFLGEKYI RRVRMRPGVA CSVSQAQKDE LILEGNDIEL VSNSAALIQQ ATTVKNKDIR KFLDGIYVSE KGTVQQADE Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L6P family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.
Especificaciones
Especificaciones
| Antígeno | RPL9 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | RPL9 |
| N.º de referencia del gen | P32969, P51410 |
| Alias de gen | 60S ribosomal protein L9; 60S ribosomal protein L9-like protein; ab02c03; fa93a01; L9; Large ribosomal subunit protein uL6; mg:ab02c03; NPC-A-16; OK/SW-cl.103; ribosomal protein L9; ribosomal protein L9-like; ribosomal protein L9-like protein; RPL9; rpl9 protein; RPL9P7; RPL9P8; RPL9P9; similar to ribosomal protein L9; wu:fa93a01; zgc:103730 |
| Símbolos de los genes | RPL9 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
