missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL38 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA568074
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Predicted to react with Human, Mouse, and Rat samples.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L38E family of ribosomal proteins. It is located in the cytoplasm. Alternative splice variants have been identified, both encoding the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome, including one located in the promoter region of the type 1 angiotensin II receptor gene.
Especificaciones
| RPL38 | |
| Polyclonal | |
| Unconjugated | |
| RPL38 | |
| 0610025G13Rik; 60S ribosomal protein L38; L38; Large ribosomal subunit protein eL38; Rbt; ribosomal protein L38; Rpl38; Ts; Tss | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6169, 67671, 689284 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 30% glycerol, 0.87% NaCl and 0.01% sodium azide; pH 7.3 | |
| P63173, P63174, Q9JJI8 | |
| RPL38 | |
| Synthesized peptide derived from human RPL38. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido