missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
RPL36AL Polyclonal Antibody for Western Blot, ELISA
Cytoplasmic ribosomes, organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein, which shares sequence similarity with yeast ribosomal protein L44, belongs to the L44E (L36AE) family of ribosomal proteins. This gene and the human gene officially named ribosomal protein L36a (RPL36A) encode nearly identical proteins; however, they are distinct genes. Although the name of this gene has been referred to as ribosomal protein L36a (RPL36A), its official name is ribosomal protein L36a-like (RPL36AL). As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Especificaciones
Especificaciones
| Antígeno | RPL36AL |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| génica | RPL36AL |
| N.º de referencia del gen | Q969Q0 |
| Alias de gen | 2410038A03Rik; 60S ribosomal protein L36a; 60S ribosomal protein L36a-like; 60S ribosomal protein L44; Large ribosomal subunit protein eL42-like; large subunit ribosomal protein L36a; MGC111574; ribosomal protein HL44; ribosomal protein L36a; ribosomal protein L36a like; ribosomal protein L36a-like; Rpl36a; RPL36A protein; RPL36AL; Rpl44 |
| Símbolos de los genes | RPL36AL |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
