missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA549959
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The antibody detects endogenous levels of total RPL36 protein.
The exosome is involved in a multitude of cellular RNA processing and degradation events. DIS3, also known as exosome complex exonuclease RRP44, is a ribonuclease that acts directly in the processing, turnover, and surveillance of a large number of distinct RNA species. DIS3 localizes to both the cytoplasm and the nucleus and contains one PINc domain. It is widely expressed with highest expression in testis and is required for processing of 7S pre-RNA into a mature nuclear complex and, ultimately, for proper mitotic progression. Abnormal expression levels of DIS3 may be associated with colon cancer, suggesting a role for DIS3 in tumorigenesis.
Especificaciones
| RPL36 | |
| Polyclonal | |
| Unconjugated | |
| Rpl36 | |
| 60S ribosomal protein L36; L36; Large ribosomal subunit protein eL36; ribosomal protein L36; RPL36 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25873 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9Y3U8 | |
| Rpl36 | |
| Synthesized peptide derived from internal of human RPL36. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido