missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL30 Polyclonal Antibody

Descripción
Peptide sequence: LKMIRQGKAK LVILANNCPA LRKSEIEYYA MLAKTGVHHY SGNNIELGTA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L30E family of ribosomal proteins. It is located in the cytoplasm. This gene is co-transcribed with the U72 small nucleolar RNA gene, which is located in its fourth intron. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Especificaciones
Especificaciones
| Antígeno | RPL30 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 2% sucrose and 0.09% sodium azide |
| génica | RPL30 |
| N.º de referencia del gen | P62888 |
| Alias de gen | 60S ribosomal protein L30; fa93d05; hypothetical protein LOC513031; L30; Large ribosomal subunit protein eL30; ribosomal protein L30; RPL30; wu:fa93d05; zgc:56640; zgc:77683 |
| Símbolos de los genes | RPL30 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?