Descripción
The RPL3 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human RPL3. This antibody recognizes human, mouse, rat antigen. The RPL3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Especificaciones
Especificaciones
| Antígeno | RPL3 |
| Aplicaciones | Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | 2G1B8 |
| Conjugado | CoraLite 488 |
| Especie del huésped | Mouse |
| Cantidad | 100 μL |
| Primario o secundario | Primary |
| ID de gen (Entrez) | 6122 |
| Especies diana | Human, Rat, Mouse |
| Mostrar más |