missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA541715
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Peptide sequence: KKEKARVITE EEKNFKAFAS LRMARANARL FGIRAKRAKE AAEQDVEKKK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. RPL13 is a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L13E family of ribosomal proteins. It is located in the cytoplasm. This gene is expressed at significantly higher levels in benign breast lesions than in breast carcinomas. Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L13E family of ribosomal proteins. It is located in the cytoplasm. This gene is expressed at significantly higher levels in benign breast lesions than in breast carcinomas. Transcript variants derived from alternative splicing and/or alternative polyadenylation exist; these variants encode the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Especificaciones
| RPL13 | |
| Polyclonal | |
| Unconjugated | |
| RPL13 | |
| 60S ribosomal protein L13; A52; BBC1; breast basic conserved protein 1; D16S444E; D16S44E; FLJ27453; FLJ27454; L13; Large ribosomal subunit protein eL13; LOC508221 protein; MGC117342; MGC71373; OK/SW-cl.46; ribosomal protein L13; RPL13; RPL13 protein-like; zgc:92272 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6137 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P26373 | |
| RPL13 | |
| Synthetic peptide directed towards the C-terminal of human RPL13 (aa 162-211). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur