missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL10 Monoclonal Antibody (OTI9A8), TrueMAB™, OriGene
Descripción
RPL10 Monoclonal antibody specifically detects RPL10 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Especificaciones
Especificaciones
| Antígeno | RPL10 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI9A8 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | DMRT1 |
| N.º de referencia del gen | Q9Y5R6 |
| Alias de gen | 24.6 kDa protein; 24.6kDa protein; 60S ribosomal protein L10; AUTSX5; D0HXS648; DXHXS648; DXHXS648E; DXS648; DXS648E; L10; Laminin receptor homolog; Large ribosomal subunit protein uL16; NOV; Protein QM; Protein QM homolog; QM; QM16; ribosomal protein 10; ribosomal protein L10; Rpl10; RPLPO; Tumor suppressor QM; Wilms tumor-related protein |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?