Learn More
RPE65 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Código de producto. 18019805 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
100 μL
20 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
18019805 100 μL 100 microlitros
18028415 20 μL 20 microlitros
2 options
Código de producto. 18019805
Marca: Proteintech CL48817939100UL
Rabbit Polyclonal Antibody
The RPE65 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RPE65. This antibody recognizes human, mouse, rat antigen. The RPE65 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Especificaciones
Antígeno RPE65
Aplicaciones Immunofluorescence
Concentración 1000 μg/mL
Conjugado CoraLite Plus 488
Símbolos de los genes LCA2, mRPE65, rd12, Retinoid isomerohydrolase, Retinol isomerase, RP20, RPE65, sRPE65
Especie del huésped Rabbit
Cantidad 100 μL
Primario o secundario Primary
ID de gen (Entrez) 6121
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Peso molecular del anticuerpo 150 kDa
Formulario Liquid
Isotype IgG