Learn More
Invitrogen™ Rotavirus NCDV Polyclonal Antibody, Biotin
Descripción
The PA1-73016 antibody reacts with Rotavirus Bovine NCDV. PA1-73016 has been successfully used in ELISA applications. The PA1-73016 antibody was raised against Rotavirus ICP + virions. Recommended short term storage of less than 6 months store at 4°C. For extended storage, store at -20°C.
Rotavirus is a double-stranded RNA virus in the family Reoviridae. Five species of rotavirus (A-E) cause disease in animals with species A being the most common in human. Within rotavirus A there are different strains, called serotypes, based on the dual classification system of two proteins on the surface of the virus. Six viral proteins (VPs), or structural proteins, form the virion and six non-structural proteins (NSPs) are expressed in infected cells. Rotavirus is the most common cause of severe diarrhoea among infants and young children. Rotavirus gastroenteritis is a mild to severe disease characterised by vomiting, watery diarrhoea, and low-grade fever, but still leads to hundreds of thousands of deaths each year in developing countries.
Especificaciones
Especificaciones
| Antígeno | Rotavirus NCDV |
| Aplicaciones | ELISA |
| Clasificación | Polyclonal |
| Concentración | 4-5 mg/mL |
| Conjugado | Biotin |
| Formulación | PBS with 0.1% sodium azide; pH 7.2 |
| Alias de gen | Reoviridae |
| Especie del huésped | Goat |
| Inmunógeno | Bovine (Nebraska Calf Diarrhea Virus). |
| Método de purificación | IgG fraction |
| Mostrar más |