missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rotavirus Capsid Monoclonal Antibody (J74D)

Descripción
The MA1-7282 antibody reacts with Rotavirus capsid in viral and infected tissue samples. MA1-7282 has been successfully used in ELISA applications. The MA1-7282 antibody was raised against purified rotavirus capsid.
Rotavirus is a double-stranded RNA virus in the family Reoviridae. Five species of rotavirus (A-E) cause disease in animals with species A being the most common in human. Within rotavirus A there are different strains, called serotypes, based on the dual classification system of two proteins on the surface of the virus. Six viral proteins (VPs), or structural proteins, form the virion and six non-structural proteins (NSPs) are expressed in infected cells. Rotavirus is the most common cause of severe diarrhoea among infants and young children. Rotavirus gastroenteritis is a mild to severe disease characterised by vomiting, watery diarrhoea, and low-grade fever, but still leads to hundreds of thousands of deaths each year in developing countries.
Especificaciones
Especificaciones
| Antígeno | Rotavirus Capsid |
| Aplicaciones | ELISA |
| Clasificación | Monoclonal |
| Clon | J74D |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.1% sodium azide; pH 7.2 |
| Alias de gen | Reoviridae |
| Especie del huésped | Mouse |
| Inmunógeno | Purified rotavirus capsid. |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?