missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies RosetteSep™ Human T Cell Enrichment Cocktail

Código de producto. 15623934
Encompass
Change view
Click to view available options
Cantidad:
200 mL de sangre completa
40 mL de sangre completa
Tamaño de la unidad:
1 set
200 ml
Código de producto. Cantidad unitSize
15623934 200 mL de sangre completa 200 ml
17197651 40 mL de sangre completa 1 set
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15623934

Marca: STEMCELL Technologies 15061

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Immunodensity isolation of untouched T cells

  • The RosetteSep™ Human T Cell Enrichment Cocktail is designed to isolate T cells from whole blood by negative selection
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-T cells and glycophorin A on red blood cells (RBCs)
  • When centrifuged over a buoyant density medium such as RosetteSep™ DM-L (Catalog #15705) or Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs
  • The purified T cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium

Specifications

Tipo de célula T Cells
Para utilizar con (aplicación) Cell Isolation
Cantidad 200 mL de sangre completa
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.