missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies RosetteSep™ Human Bone Marrow Progenitor Cell Pre-Enrichment Cocktail

Código de producto. 17157661
Encompass
Change view
Click to view available options
Cantidad:
200 mL de médula ósea
40 mL de médula ósea
Tamaño de la unidad:
1 set
Código de producto. Cantidad unitSize
17157661 40 mL de médula ósea 1 set
17127671 200 mL de médula ósea 1 set
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Product Code. 17157661

missing translation for 'mfr': STEMCELL Technologies 15027

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Immunodensity pre-enrichment of untouched stem/progenitor cells from bone marrow

  • The RosetteSep™ Human Bone Marrow Progenitor Cell Pre-Enrichment Cocktail is designed to pre-enrich progenitor cells by removing lineage-specific cells from whole bone marrow by negative selection
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD3, CD11b, CD14, CD16, CD19, CD56, CD66b and glycophorin A on red blood cells (RBCs)
  • When centrifuged over a buoyant density medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs
  • The pre-enriched progenitor cells are present as an enriched population at the interface between the plasma and the buoyant density medium

Specifications

Tipo de célula Hematopoietic Stem and Progenitor Cells
Para utilizar con (aplicación) Cell Isolation
Cantidad 40 mL de médula ósea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.