Learn More
STEMCELL Technologies RosetteSep™ Human B Cell Enrichment Cocktail
Código de producto. 17187661
Encompass
Change view
Click to view available options
Cantidad:
200 mL de sangre completa
40 mL de sangre completa
Tamaño de la unidad:
1 set
17187661 200 mL de sangre completa 1 set
17127661 40 mL de sangre completa 1 set
2 options
Código de producto. 17187661
Marca: STEMCELL Technologies 15064
Immunodensity isolation of untouched B cells
  • The RosetteSep™ Human B Cell Enrichment Cocktail is designed to isolate B cells from whole blood by negative selection
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-B cells and glycophorin A on red blood cells (RBCs)
  • When centrifuged over a buoyant density medium such as RosetteSep™ DM-L (Catalog #15705) or Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs
  • The purified B cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium
Especificaciones
Tipo de célula B Cells
Para utilizar con (aplicación) Cell Isolation
Cantidad 200 mL de sangre completa