Learn More
STEMCELL Technologies RosetteSep™ HLA T Cell Enrichment Cocktail
Descripción
- The RosetteSep™ HLA T Cell Enrichment Cocktail is designed to isolate T cells from whole blood by negative selection.
- Unwanted cells are targeted for removal with Tetrameric Antibody Complexes (TAC) recognizing non-T cells and red blood cells (RBCs).
- When centrifuged over a buoyant density medium such as RosetteSep™ DM-L (Catalog #15705) or Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs.
- The purified T cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium.
- The desired cells are immediately ready for serology or flow cytometry crossmatch assays, or other downstream applications.
Note: In light of the new EU Regulation 2017/746 (IVDR) of the European Parliament and of the Council on in vitro diagnostic medical devices, which is expected to become effective in 2022, STEMCELL recently deregistered and removed the CE IVD claims associated with these products, and they are now labeled For Research Use Only. Please note there have been no changes to the form, function and quality assurances associated with these products.
Especificaciones
Especificaciones
| Tipo de célula | T Cells |
| Para utilizar con (aplicación) | Cell Isolation; In Vitro Diagnostic |
| Cantidad | 250 mL de sangre completa |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.