Learn More
STEMCELL Technologies RosetteSep™ CTC Enrichment Cocktail Containing Anti-CD36
Código de producto. 17197671
Encompass
Change view
Click to view available options
Cantidad:
200 mL de sangre completa
40 mL de sangre completa
Tamaño de la unidad:
1 set
17197671 40 mL de sangre completa 1 set
17157681 200 mL de sangre completa 1 set
2 options
Código de producto. 17197671
Marca: STEMCELL Technologies 15127
Immunodensity isolation of untouched circulating tumor cells
  • The RosetteSep™ CTC Enrichment Cocktail Containing Anti-CD36 is designed to enrich circulating epithelial tumor cells from fresh whole blood by negative selection
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD2, CD16, CD19, CD36, CD38, CD45, CD66b and glycophorin A on red blood cells (RBCs)
  • When centrifuged over a density gradient medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs
  • The purified tumor cells are present as a highly enriched population at the interface between the plasma and the density gradient medium
  • This cocktail is compatible with small-cell carcinoma samples, including lung cancer samples
  • If you are working with breast cancer samples, use the RosetteSep™ CTC Enrichment Cocktail Containing Anti-CD56 (Catalog #15137)
Especificaciones
Tipo de célula Cancer Cells and Cell Lines
Para utilizar con (aplicación) Cell Isolation
Cantidad 40 mL de sangre completa