Learn More
STEMCELL Technologies RoboSep™ Human Myeloid DC Enrichment Kit
Código de producto. 17187901
Encompass
Change view
Click to view available options
Cantidad:
2 x 109 celdas
Tamaño de la unidad:
1 set
17187901 2 x 109 celdas 1 set
1 options
Código de producto. 17187901
Marca: STEMCELL Technologies 19061RF
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
  • The EasySep™ Human Myeloid DC Enrichment Kit is designed to isolate myeloid dendritic cells (mDC) from fresh peripheral blood mononuclear cells or ammonium chloride-lysed leukapheresis by negative selection
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-mDCs and dextran-coated magnetic particles
  • The labeled cells are separated using the EasySep™ magnet without the use of columns
  • Desired cells are poured off into a new tube
Especificaciones
Tipo de célula Dendritic Cells
Para utilizar con (aplicación) Cell Isolation
Cantidad 2 x 109 celdas