Learn More
STEMCELL Technologies RoboSep™ HLA Chimerism Whole Blood CD4 Positive Selection Kit
Código de producto. 17187801
Encompass
Change view
Click to view available options
Cantidad:
60 mL de sangre completa
Tamaño de la unidad:
1 set
17187801 60 mL de sangre completa 1 set
1 options
Código de producto. 17187801
Marca: STEMCELL Technologies 17888RF
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
  • The EasySep™ HLA Chimerism Whole Blood CD4 Positive Selection is designed to isolate CD4+ cells from fresh whole blood or buffy coat by positive selection
  • Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD4 and dextran-coated magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • The CD4 antigen is expressed on helper T cells and more weakly on monocytes and tissue macrophages
  • This product replaces the EasySep™ Human Whole Blood CD4 Positive Selection Kit (Catalog #18082) for even faster cell isolations
Especificaciones
Tipo de célula T Cells, CD4+
Para utilizar con (aplicación) Cell Isolation
Cantidad 60 mL de sangre completa