Learn More
STEMCELL Technologies RoboSep™ HLA Chimerism Whole Blood CD33 Positive Selection Kit
Código de producto. 17127801
Encompass
Change view
Click to view available options
Cantidad:
1 x 109 Celdas
Tamaño de la unidad:
1 set
17127801 1 x 109 Celdas 1 set
1 options
Código de producto. 17127801
Marca: STEMCELL Technologies 17885RF
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
  • The EasySep™ HLA Chimerism Whole Blood CD33 Positive Selection Kit is designed to isolate highly purified myeloid (including CD14+ and CD66b+) cells from fresh whole blood by positive selection
  • Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD33 and dextran-coated magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • The CD33 antigen is expressed on myeloid progenitor cells, granulocytes and monocytes
  • This product replaces the EasySep™ Human Whole Blood CD33 Positive Selection Kit (Catalog #18287) and EasySep™ HLA Whole Blood CD33 Positive Selection Kit (Catalog #18287HLA) for even faster cell isolations
Especificaciones
Tipo de célula Granulocytes and Subsets, Myeloid Cells
Para utilizar con (aplicación) Cell Isolation
Cantidad 1 x 109 Celdas