Learn More
STEMCELL Technologies RoboSep™ HLA Chimerism Whole Blood CD15 Positive Selection Kit
Código de producto. 17147791
Encompass
Change view
Click to view available options
Cantidad:
60 mL de capa leucocitaria o sangre completa
Tamaño de la unidad:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
17147791 60 mL de capa leucocitaria o sangre completa 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Código de producto. 17147791 Proveedor STEMCELL Technologies N.º de proveedor 17881RF
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
  • The EasySep™ HLA Chimerism Whole Blood CD15 Positive Selection Kit is designed to isolate highly purified CD15+ cells from fresh whole blood by positive selection
  • Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD15 and dextran-coated magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • The CD15 antigen is expressed on granulocytes and monocytes
  • This product replaces the EasySep™ Human Whole Blood CD15 Positive Selection Kit (Catalog #18681) and EasySep™ HLA Whole Blood CD15 Positive Selection Kit (Catalog #18681HLA), for even faster cell isolations
Especificaciones
Tipo de célula Myeloid Cells
Para utilizar con (aplicación) Cell Isolation
Cantidad 60 mL de capa leucocitaria o sangre completa