missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ROBO3 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 30019819
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
30019819 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30019819

Marca: Invitrogen™ PA5148420

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

ROBO3 Polyclonal Antibody for IHC (P)

This gene is a member of the Roundabout (ROBO) gene family that controls neurite outgrowth, growth cone guidance, and axon fasciculation. ROBO proteins are a subfamily of the immunoglobulin transmembrane receptor superfamily. SLIT proteins 1-3, a family of secreted chemorepellants, are ligands for ROBO proteins and SLIT/ROBO interactions regulate myogenesis, leukocyte migration, kidney morphogenesis, angiogenesis, and vasculogenesis in addition to neurogenesis. This gene, ROBO3, has a putative extracellular domain with five immunoglobulin (Ig)-like loops and three fibronectin (Fn) type III motifs, a transmembrane segment, and a cytoplasmic tail with three conserved signaling motifs: CC0, CC2, and CC3 (CC for conserved cytoplasmic). Unlike other ROBO family members, ROBO3 lacks motif CC1. The ROBO3 gene regulates axonal navigation at the ventral midline of the neural tube. In mouse, loss of Robo3 results in a complete failure of commissural axons to cross the midline throughout the spinal cord and the hindbrain. Mutations ROBO3 result in horizontal gaze palsy with progressive scoliosis (HGPPS); an autosomal recessive disorder characterized by congenital absence of horizontal gaze, progressive scoliosis, and failure of the corticospinal and somatosensory axon tracts to cross the midline in the medulla.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno ROBO3
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 100 μg/mL
Conjugado Unconjugated
Formulación PBS with 0.03% ProClin 300; pH 7
génica Robo3
N.º de referencia del gen Q96MS0
Alias de gen FLJ21044; HGPPS; HGPS; RBIG1; retinoblastoma inhibiting gene 1; retinoblastoma-inhibiting gene 1 protein; Rig1; Rig-1; ROBO3; Robo3a; Robo3b; roundabout guidance receptor 3; roundabout homolog 3; roundabout homolog 3 (Drosophila); roundabout, axon guidance receptor, homolog 3; roundabout, axon guidance receptor, homolog 3 (Drosophila); roundabout-like protein 3
Símbolos de los genes Robo3
Especie del huésped Rabbit
Inmunógeno E. coli-derived Human Robo3 fragment.
Método de purificación Antigen affinity chromatography, Protein A
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 64221
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.