missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ro 61-8048, MedChemExpress
Descripción
Ro 61-8048 is an orally active and selective inhibitor of kynurenine 3-hydroxylase, with an IC50 of 37 nM. Ro 61-8048 provokes a significant increase of extracellular kynurenic acid concentrations.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Ro 61-8048 |
| CAS | 199666-03-0 |
| Color | Light Yellow |
| Notas de grado de pureza | Research |
| Fórmula molecular | C17H15N3O6S2 |
| Cantidad | 5 mg |
| Información de solubilidad | DMSO : ≥ 59 mg/mL (139.99 mM) |
| SMILES | O=S(C1=CC=C(OC)C(OC)=C1)(NC2=NC(C3=CC=CC([N+]([O-])=O)=C3)=CS2)=O |
| Peso molecular (g/mol) | 421.45 |
| Formula Weight (peso de la fórmula) | 421.45 |
| Mostrar más |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?