missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF125 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13290-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TRAC-1 is a 232 amino acid protein containing an N-terminal RING-type zinc finger domain. It functions as a positive regulator in the T-cell receptor signaling pathway, thus leading to T-cell activation. It mediates ubiquitination and subsequent proteasomal degradation of target proteins. TRAC-1 is expressed in lymphoid tissues, including bone marrow, spleen, thymus as well as in other tissues. It is predomitly expressed in the CD4+ and CD8+ T-cells, suggesting that it is preferentially confined to T-cells.Especificaciones
| RNF125 | |
| Polyclonal | |
| Unconjugated | |
| RNF125 | |
| ring finger protein 125, RNF125, TRAC 1, TRAC1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 361296, 54941, 67664 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.52 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96EQ8, Q9D9R0 | |
| Rnf125 | |
| RNF125 Fusion Protein Ag3908 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido