missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ripertamab Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
Marca: Invitrogen™ MA559181
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Ripertamab, also known as SCT400, is a recombinant chimeric human-mouse anti-CD20 monoclonal antibody. The CD20 gene, also known as MS4A1, encodes a B cell-specific membrane-bound protein that plays a crucial role in B cell development and function. Located on chromosome 11q12 in humans, the MS4A1 gene spans 16 kb and includes eight exons. CD20 is a part of the membrane-spanning 4-domain family, subfamily A (MS4A), which encompasses 18 tetraspan proteins. The prominent CD20 mRNA variant is 2.8 kb long, although other shorter variants exist. This phosphoprotein, approximately 33-37 kDa, is expressed on the surface of almost all normal and malignant B cells, making it an ideal target for therapeutic antibodies like rituximab. CD20 is known to interact with several proteins such as CD40, MHCII, and CD81, playing a role in B cell receptor signaling and other immune processes. Despite its widespread clinical use in B cell depletion therapies, the precise biological function of CD20 remains partly unresolved.
Especificaciones
| Ripertamab Chimeric | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM Histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido