missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RIP3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Marca:  Invitrogen™ PA5119682

Código de producto. 17751084

  • 514.00€ / 100 microlitros

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

Positive Control: MEF cell lysates. Subcellular Location: Mitochondrion, Cytosol, Plasma membrane.

RIPK3 is a member of the receptor-interacting protein (RIP) family of serine/threonine protein kinases, which contains a C-terminal domain unique from other RIP family members. RIPK3 is predominantly localized to the cytoplasm, and specifically, are colocalized in the mitochondrion which can undergo nucleocytoplasmic shuttling dependent on novel nuclear localization and export signals. RIPK3 is a component of the tumor necrosis factor (TNF) receptor-I signaling complex, and can induce apoptosis and weakly activate the NF-kappaB transcription factor and caspase-8 prevents RIPK3-dependent necrosis without inducing apoptosis by functioning in a proteolytically active complex with CFLAR and that this complex is required for the protective function.
TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

RIP3
Polyclonal
Unconjugated
RIPK3
2610528K09Rik; AW107945; Hcyp2; Homocysteine respondent protein HCYP2; mRIP3; receptor interacting protein 3; receptor interacting serine/threonine kinase 3; receptor-interacting protein 3; receptor-interacting serine/threonine-protein kinase 3; receptor-interacting serine-threonine kinase 3; Rip3; RIP-3; Ripk3; RIP-like protein kinase 3
Rabbit
Antigen affinity chromatography
RUO
246240, 56532
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Q9QZL0, Q9Z2P5
RIPK3
Synthetic peptide within mouse RIP3 aa 440-486.
100 μL
Primary
Mouse, Rat
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Promociones

Promociones

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.