missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RIP2 Polyclonal Antibody

Descripción
RIP2 Polyclonal Antibody for Western Blot, IHC (P)
RIPK2 is a serine/threonine kinase that is involved in cell activation, proliferation, differentiation, and apoptosis. Upon stimulation by bacterial peptidoglycans, NOD1 and NOD2 are activated, oligomerize and recruit RIPK2 through CARD-CARD domains. Contributes to the tyrosine phosphorylation of the guanine exchange factor ARHGEF2 through Src tyrosine kinase leading to NF-kappaB activation by NOD2. Once recruited, RIPK2 autophosphorylates and undergoes 'Lys-63'-linked polyubiquitination by E3 ubiquitin ligases XIAP, BIRC2 and BIRC3. The polyubiquitinated protein mediates the recruitment of MAP3K7/TAK1 to IKBKG/NEMO and induces 'Lys-63'-linked polyubiquitination of IKBKG/NEMO and subsequent activation of IKBKB/IKKB. In turn, NF-kappa-B is released from NF-kappa-B inhibitors and translocates into the nucleus where it activates the transcription of hundreds of genes involved in immune response, growth control, or protection against apoptosis. Plays also a role during engagement of the T-cell receptor (TCR) in promoting BCL10 phosphorylation and subsequent NF-kappa-B activation.
Especificaciones
Especificaciones
| Antígeno | RIP2 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.09% sodium azide; pH 7.4 |
| génica | RIPK2 |
| N.º de referencia del gen | O43353, P58801 |
| Alias de gen | 2210420D18Rik; CARD3; CARD-carrying kinase; CARD-containing IL-1 beta ICE-kinase; CARD-containing interleukin-1 beta-converting enzyme (ICE)-associated kinase; CARD-containing interleukin-1 beta-converting enzyme-associated kinase; CARDIAK; CCK; D4Bwg0615e; EC 2.7.11.1; GIG30; growth-in; growth-inhibiting gene 30; kinase RIPK2; receptor (TNFRSF)-interacting serine-threonine kinase 2; receptor interacting serine/threonine kinase 2; receptor-interacting protein (RIP)-like interacting caspase-like apoptosis regulatory protein (CLARP) kinase; receptor-interacting protein 2; receptor-interacting serine/threonine-protein kinase 2; receptor-interacting serine-threonine kinase 2; RICK; RIP 2; RIP2; RIP-2; Ripk2; RIP-like; RIP-like-interacting CLARP kinase; Tyrosine-protein kinase RIPK2; UNQ277/PRO314/PRO34092; WUGSC:H_RG437L15.1 |
| Símbolos de los genes | RIPK2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?