Learn More
Epredia™ Dehydrant Alcohol
Dehydrant Alcohol products are specifically designed for tissue processing and staining of histological and cytological specimens. Available in 70%, 80%, 95% and 100% blends for complete dehydration.
Descripción
Descripción
  • Blend of ethyl, methyl and isopropyl alcohols
  • Can be used as a dye or stain solvent
  • Miscible with all clearing agents